ENA161209308 Enamine

methyl 4-(3-chlorophenyl)-2,4-dioxobutanoate

  • MDL number MFCD10478941
  • Cas Number : 501653-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29...31
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27175

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-27175
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart