ENA161209313 Enamine

3-morpholin-4-yl-3-oxopropan-1-amine hydrochloride

  • MDL number MFCD07366749
  • Cas Number : 173336-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Salt Data HCl
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27314

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27314
Availability details for
Quantity
  • Availability
  • Add to Cart