ENA161209318 Enamine

1-(prop-2-yn-1-yl)piperazine dihydrochloride

  • MDL number MFCD00039029
  • Cas Number : 90000-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27346
Availability details for
Quantity
  • Availability
  • Add to Cart