ENA161209323 Enamine

4-[(propan-2-yl)amino]butanoic acid hydrochloride

  • MDL number MFCD09373417
  • Cas Number : 1094598-19-2
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27445
Availability details for
Quantity
  • Availability
  • Add to Cart