ENA161209329 Enamine

methyl 2,3-dihydro-1H-indole-5-carboxylate

  • MDL number MFCD07371630
  • Cas Number : 141452-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27629
Availability details for
Quantity
  • Availability
  • Add to Cart