ENA161209338 Enamine

N-(2-aminophenyl)-N-(4-methoxyphenyl)amine hydrochloride

  • MDL number MFCD07700776
  • Cas Number : 1184991-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02791

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02791
Availability details for
Quantity
  • Availability
  • Add to Cart