ENA161209352 Enamine

N-(2-aminoethyl)furan-2-carboxamide hydrochloride

  • MDL number MFCD02675831
  • Cas Number : 81253-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28769
Availability details for
Quantity
  • Availability
  • Add to Cart