ENA161209367 Enamine

2-[4-(chloromethyl)-1,3-thiazol-2-yl]-N-(2-methylphenyl)acetamide hydrochloride

  • MDL number MFCD08447159
  • Cas Number : 1171526-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07077

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07077
Availability details for
Quantity
  • Availability
  • Add to Cart