ENA161209370 Enamine

ethyl 2-amino-2-[1-(4-fluorophenyl)-3,5-dimethyl-1H-pyrazol-4-yl]acetate hydrochloride

  • MDL number MFCD09834104
  • Cas Number : 1078161-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29495
Availability details for
Quantity
  • Availability
  • Add to Cart