ENA161209378 Enamine

N-(2-aminoethyl)benzenesulfonamide hydrochloride

  • MDL number MFCD09834110
  • Cas Number : 53672-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29647

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29647
Availability details for
Quantity
  • Availability
  • Add to Cart