ENA161209386 Enamine

5-amino-4-(trifluoromethyl)pentanoic acid hydrochloride

  • MDL number MFCD00723867
  • Cas Number : 1052404-68-8
  • Molecular Weight : 221.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(aminomethyl)-5,5,5-trifluoropentanoic acid hydrochloride
Inchi Code 1S/C6H10F3NO2.ClH/c7-6(8,9)4(3-10)1-2-5(11)12;/h4H,1-3,10H2,(H,11,12);1H
Inchi Key IUYUKQUTNGOQHL-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07164
Availability details for
Quantity
  • Availability
  • Add to Cart