ENA161209397 Enamine

3-(4-aminobutyl)-1,3-thiazolidine-2,4-dione hydrochloride

  • MDL number MFCD03037692
  • Cas Number : 25565-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-33644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33644
Availability details for
Quantity
  • Availability
  • Add to Cart