ENA161209403 Enamine

1-[3-(trifluoromethyl)phenyl]-1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxylic acid

  • MDL number MFCD09049514
  • Cas Number : 926254-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35459
Availability details for
Quantity
  • Availability
  • Add to Cart