ENA161209409 Enamine

1-[5-(2-fluorophenyl)thiophen-2-yl]ethan-1-one

  • MDL number MFCD06802793
  • Cas Number : 893741-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35562
Availability details for
Quantity
  • Availability
  • Add to Cart