ENA161209412 Enamine

5-oxo-1-(2,2,2-trifluoroethyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD09944063
  • Cas Number : 1016823-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35608
Availability details for
Quantity
  • Availability
  • Add to Cart