ENA161209515 Enamine

N-[3-(aminomethyl)phenyl]furan-2-carboxamide hydrochloride

  • MDL number MFCD10686762
  • Cas Number : 1171521-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35784
Availability details for
Quantity
  • Availability
  • Add to Cart