ENA161209516 Enamine

N-({3-methylimidazo[1,5-a]pyridin-1-yl}methylidene)hydroxylamine

  • Cas Number : 1050910-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35796
Availability details for
Quantity
  • Availability
  • Add to Cart