ENA161209517 Enamine

6-chloro-2-oxo-1,2,3,4-tetrahydroquinoline-7-sulfonyl chloride

  • MDL number MFCD10686776
  • Cas Number : 1050910-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35820
Availability details for
Quantity
  • Availability
  • Add to Cart