ENA161209523 Enamine

N-{[4-(dimethylamino)phenyl]methyl}-1H-imidazole-1-carboxamide

  • MDL number MFCD10686783
  • Cas Number : 1050910-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35857
Availability details for
Quantity
  • Availability
  • Add to Cart