ENA161209524 Enamine

methyl 1,6-dimethyl-3-oxo-1H,2H,3H-pyrazolo[3,4-b]pyridine-4-carboxylate

  • MDL number MFCD10686785
  • Cas Number : 71290-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-35859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35859
Availability details for
Quantity
  • Availability
  • Add to Cart