ENA161209530 Enamine

N-[(oxolan-3-yl)methylidene]hydroxylamine

  • Cas Number : 1809056-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35890
Availability details for
Quantity
  • Availability
  • Add to Cart