ENA161209538 Enamine

3-amino-1-phenyl-2-[(pyrrolidin-1-yl)methyl]propan-1-ol

  • MDL number MFCD10686808
  • Cas Number : 1049606-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35968
Availability details for
Quantity
  • Availability
  • Add to Cart