ENA161209542 Enamine

2-amino-1-phenyl-4,5,6,7-tetrahydro-1H-indole-3-carbonitrile

  • MDL number MFCD10686813
  • Cas Number : 55817-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35983
Availability details for
Quantity
  • Availability
  • Add to Cart