ENA161209545 Enamine

3-methylcyclohexan-1-amine hydrochloride

  • MDL number MFCD10686816
  • Cas Number : 89854-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-175
Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-35997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35997
Availability details for
Quantity
  • Availability
  • Add to Cart