ENA161209546 Enamine

2-methyl-4-morpholin-4-ylaniline

  • MDL number MFCD10686817
  • Cas Number : 581-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36000

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-36000
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart