ENA161209549 Enamine

1-(4-bromophenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD10686820
  • Cas Number : 944901-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-140
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36011
Availability details for
Quantity
  • Availability
  • Add to Cart