ENA161209552 Enamine

7-ethyl-[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD10686824
  • Cas Number : 1049606-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36018
Availability details for
Quantity
  • Availability
  • Add to Cart