ENA161209568 Enamine

1-(3-bromophenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD10686843
  • Cas Number : 944905-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36116
Availability details for
Quantity
  • Availability
  • Add to Cart