ENA161209573 Enamine

methyl 5-acetyl-3-methylthieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD10686851
  • Cas Number : 1050885-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36157
Availability details for
Quantity
  • Availability
  • Add to Cart