ENA161209574 Enamine

3-(2-bromophenyl)-2-[(dimethylamino)methyl]-3-hydroxypropanenitrile

  • MDL number MFCD10686854
  • Cas Number : 1049606-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36170
Availability details for
Quantity
  • Availability
  • Add to Cart