ENA161209577 Enamine

N-[1-(5-methylfuran-2-yl)propylidene]hydroxylamine

  • Cas Number : 133712-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36181
Availability details for
Quantity
  • Availability
  • Add to Cart