ENA161209585 Enamine

1-{3-[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]propyl}piperazine dihydrochloride

  • MDL number MFCD10686871
  • Cas Number : 1170167-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36242
Availability details for
Quantity
  • Availability
  • Add to Cart