ENA161209605 Enamine

3-(3,4-difluorobenzyl)azetidine

  • MDL number MFCD09053212
  • Cas Number : 937609-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35930

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35930
Availability details for
Quantity
  • Availability
  • Add to Cart