ENA161209609 Enamine

N-(3-aminophenyl)benzamide

  • MDL number MFCD00071750
  • Cas Number : 16091-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35944

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35944
Availability details for
Quantity
  • Availability
  • Add to Cart