ENA161209612 Enamine

3-(4-methyl-2-phenylpiperazin-1-yl)propan-1-amine

  • MDL number MFCD09806291
  • Cas Number : 1016701-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35947
Availability details for
Quantity
  • Availability
  • Add to Cart