ENA161209617 Enamine

2-[(pyridin-4-yl)methoxy]benzaldehyde

  • MDL number MFCD08442137
  • Cas Number : 54402-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35963
Availability details for
Quantity
  • Availability
  • Add to Cart