ENA161209618 Enamine

1-(3,5-dichlorophenyl)-2-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD09815849
  • Cas Number : 1017427-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35964
Availability details for
Quantity
  • Availability
  • Add to Cart