ENA161209622 Enamine

2-(4-aminoquinazolin-2-yl)acetonitrile

  • MDL number MFCD10686810
  • Cas Number : 868760-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35970
Availability details for
Quantity
  • Availability
  • Add to Cart