ENA161209628 Enamine

2-(cyclopentylthio)aniline

  • MDL number MFCD09939963
  • Cas Number : 77053-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35990

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35990
Availability details for
Quantity
  • Availability
  • Add to Cart