ENA161209635 Enamine

4-(pyridine-4-amido)benzoic acid

  • MDL number MFCD00690329
  • Cas Number : 5651-66-1
  • Molecular Weight : 242.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-35854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35854
Availability details for
Quantity
  • Availability
  • Add to Cart