ENA161209643 Enamine

2-(3-methyl-5-oxo-4,5-dihydro-1,2-oxazol-4-yl)acetic acid

  • MDL number MFCD10686784
  • Cas Number : 1042541-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35858
Availability details for
Quantity
  • Availability
  • Add to Cart