ENA161209649 Enamine

2-oxo-1,3-oxazolidine-3-sulfonyl chloride

  • MDL number MFCD10686775
  • Cas Number : 710286-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-90
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35819
Availability details for
Quantity
  • Availability
  • Add to Cart