ENA161209661 Enamine

4-[(2-methylpropyl)sulfamoyl]benzoic acid

  • MDL number MFCD04035129
  • Cas Number : 10252-68-3
  • Molecular Weight : 257.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36037
Availability details for
Quantity
  • Availability
  • Add to Cart