ENA161209662 Enamine

7-amino-6-chloro-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD08444635
  • Cas Number : 40401-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36039
Availability details for
Quantity
  • Availability
  • Add to Cart