ENA161209669 Enamine

N,N-dipropylsulfamoyl chloride

  • MDL number MFCD08444784
  • Cas Number : 35877-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36048
Availability details for
Quantity
  • Availability
  • Add to Cart