ENA161209671 Enamine

N-[1-(1H-pyrazol-5-yl)ethylidene]hydroxylamine

  • Cas Number : 1192585-61-7
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35870
Availability details for
Quantity
  • Availability
  • Add to Cart