ENA161209674 Enamine

1-(4-methylphenyl)-5-(propan-2-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD10686832
  • Cas Number : 1003750-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36054
Availability details for
Quantity
  • Availability
  • Add to Cart