ENA161209675 Enamine

1-(2-methylphenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD08446377
  • Cas Number : 948293-82-1
  • Molecular Weight : 270.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36055
Availability details for
Quantity
  • Availability
  • Add to Cart