ENA161209688 Enamine

1-[4-(methoxymethyl)phenyl]methanamine

  • MDL number MFCD09737613
  • Cas Number : 132719-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36071

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-36071
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart