ENA161209691 Enamine

4-[(oxolan-2-yl)methoxy]benzamide

  • MDL number MFCD07295846
  • Cas Number : 852163-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36074
Availability details for
Quantity
  • Availability
  • Add to Cart